
Spironolactone Related Compound A

Synonym(s):
Canrenone, (17α)-17-Hydroxy-3-oxopregna-4,6-diene-21-carboxylic acid γ-lactone, NSC 261713
Empirical Formula (Hill Notation):
C22H28O3
CAS Number:
Molecular Weight:
340.46
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2