
Sodium dichromate dihydrate

Synonym(s):
Sodium bichromate, Sodium dichromate dihydrate
Linear Formula:
Na2Cr2O7 · 2H2O
CAS Number:
Molecular Weight:
298.00
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing