
Sesamolin

Synonym(s):
5-[(1S,3aR,4R,6aR)-4-(1,3-Benzodioxol-5-yloxy)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-1,3-benzodioxole
Empirical Formula (Hill Notation):
C20H18O7
CAS Number:
Molecular Weight:
370.35
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing