Skip to Content
Merck
(S)-(+)-Ketoprofen 99%

(S)-(+)-Ketoprofen

(S)-(+)-Ketoprofen 99%
Synonym(s):
(S)-(+)-3-Benzoyl-α-methylbenzeneacetic acid, (S)-2-(3-Benzoylphenyl)propionic acid
Linear Formula:
C6H5COC6H4CH(CH3)CO2H
CAS Number:
Molecular Weight:
254.28

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing