
Scoparone

Synonym(s):
6,7-Dimethoxycoumarin, 6,7-Dimethylesculetin, O-Methylisoscopoletin, O-Methylscopoletin, Aesculetin dimethyl ether, Escoparone, Esculetin 6,7-dimethyl ether, Scoparone, Scopoletin methyl ether
Empirical Formula (Hill Notation):
C11H10O4
CAS Number:
Molecular Weight:
206.19
Products
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing