![Sanguinarine chloride phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/309/570/716c81e9-c765-4c26-bae8-b41a507ab080/640/716c81e9-c765-4c26-bae8-b41a507ab080.png)
Sanguinarine chloride
![Sanguinarine chloride phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/309/570/716c81e9-c765-4c26-bae8-b41a507ab080/640/716c81e9-c765-4c26-bae8-b41a507ab080.png)
Empirical Formula (Hill Notation):
C20H14ClNO4
CAS Number:
Molecular Weight:
367.78
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing