
Rosuvastatin Calcium

Synonym(s):
Rosuvastatin calcium salt
Empirical Formula (Hill Notation):
C44H54CaF2N6O12S2
CAS Number:
Molecular Weight:
1001.14
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2