
Ribavirin Related Compound A

Synonym(s):
1-β-D-Ribofuranosyl-1H-1,2,4-triazole-3-carboxylic acid
Empirical Formula (Hill Notation):
C8H11N3O6
CAS Number:
Molecular Weight:
245.19
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing