
Quinapril Related Compound A

Synonym(s):
Quinapril diketopiperazine, (αS,3S,11aS)-1,3,4,6,11,11a-Hexahydro-3-methyl-1,4-dioxo-α-(2-phenylethyl)-2H-pyrazino[1,2-b]isoquinoline-2-acetic acid ethyl ester
Empirical Formula (Hill Notation):
C25H28N2O4
CAS Number:
Molecular Weight:
420.50
Products
Brand
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2