
Pyridine hydrobromide

Synonym(s):
Pyridinium bromide
Empirical Formula (Hill Notation):
C5H5N · HBr
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing