
Pristimerin

Synonym(s):
(9b,13a,14b,20a)-3-Hydroxy-9,13-dimethyl-2-oxo-24,25,26 -trinoroleana-1(10),3,5,7-tertraen-29-oic acid methyl ester, Celastrol methyl ester, Pristimerin
Empirical Formula (Hill Notation):
C30H40O4
CAS Number:
Molecular Weight:
464.64
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Reference Material Type
Special Grade
Showing 1-3 of 3