Skip to Content
Merck
Poly(acrylonitrile-co-butadiene), dicarboxy terminated average Mn ~3,800, acrylonitrile 8-12 wt. %

Poly(acrylonitrile-co-butadiene), dicarboxy terminated

Poly(acrylonitrile-co-butadiene), dicarboxy terminated average Mn ~3,800, acrylonitrile 8-12 wt. %
Linear Formula:
HO2C[CH2CH(CN)]x(CH2CH=CHCH2)yCO2H
CAS Number:

Products

Brand
Physical Form
Chemical Composition
Polymer Composition
Polymer Type
Showing 1-1 of 1