
Piperine

Synonym(s):
1-Piperoylpiperidine, Piperine
Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing