
Picrocrocin

Synonym(s):
(R)-4-(β-D-Glucopyranosyloxy)-β-cyclocitral, Pikrocrocin, Saffron-bitter
Empirical Formula (Hill Notation):
C16H26O7
CAS Number:
Molecular Weight:
330.37
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing