
Phthalimide

Synonym(s):
1,3-Dihydro-1,3-dioxoisoindole, Phthalimide
Empirical Formula (Hill Notation):
C8H5NO2
CAS Number:
Molecular Weight:
147.13
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing