
Oxybutynin hydrochloride for system suitability

Synonym(s):
Oxybutynin chloride, α-Phenylcyclohexaneglycolic acid 4-(diethylamino)-2-butynyl ester hydrochloride
Empirical Formula (Hill Notation):
C22H31NO3 · HCl
CAS Number:
Molecular Weight:
393.95
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing