
O-(tert-Butyldimethylsilyl)hydroxylamine

Synonym(s):
Aminoxy-tert-butyldimethylsilane
Linear Formula:
(CH3)3CSi(CH3)2ONH2
CAS Number:
Molecular Weight:
147.29
Products
Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing