
Orlistat Related Compound A

Synonym(s):
(3S,4S)-3-Hexyl-4-[(2R)-2-hydroxytridecyl]-2-oxetanone
Empirical Formula (Hill Notation):
C22H42O3
CAS Number:
Molecular Weight:
354.57
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2