
O-(4-Nitrophenylphosphoryl)choline

Synonym(s):
p-Nitrophenylphosphorylcholine, NPPC, Phosphocholine 4-nitrophenyl ester
Empirical Formula (Hill Notation):
C11H17N2O6P
CAS Number:
Molecular Weight:
304.24
Products
Product Category
Brand
Formula Weight
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing