
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride

Synonym(s):
PFBHA·HCl
Linear Formula:
C6F5CH2ONH2·HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique
Showing 1-2 of 2