Skip to Content
BG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel silver
Products
Manufacturer
Physical Form
Format
Length (mm)
OD (mm)
Quality Segment
Reference Material Type
Special Grade
certified reference material (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF53077259
Nickel silver,
rod, Cu 45%/Zn 43%/Ni 10%/Pb 2%, 20.5 mm diameter, length 100 mm
Expand
View Pricing
GF70065547
Nickel silver,
tube, Cu62%/Ni18%/Zn20%, outside diameter 1.1 mm, length 970 mm, wall thickness 0.21 mm
Expand
View Pricing
GF38456545
Nickel silver,
rod, Cu 45%/Zn 43%/Ni 10%/Pb 2%, 20.5 mm diameter, length 500 mm
Expand
View Pricing
GF63666737
Nickel silver,
rod, Cu45%/Zn43%/Ni10%/Pb2%, 20.5 mm diameter, length 200 mm
Expand
View Pricing
GF86636593
Nickel silver,
rod, Cu45%/Zn43%/Ni10%/Pb2%, 20.5 mm diameter, length 1000 mm
Expand
View Pricing
NIST880
Nickel silver,
NIST
®
SRM
®
880, (CDA 770)
Expand
View Pricing