
Naproxen Impurity A

Synonym(s):
O-Desmethylnaproxen, (S)-6-Hydroxy-α-methyl-2-naphthaleneacetic acid, d-2-(6-Hydroxy-2-naphthyl)proprionic acid
Empirical Formula (Hill Notation):
C13H12O3
CAS Number:
Molecular Weight:
216.23
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing