
Monoclonal Anti-CD3 antibody produced in mouse

Synonym(s):
Monoclonal Anti-CD3, Anti-CD3 (activation epitope)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing