
Melamine-(triamine-15N3)

Synonym(s):
1,3,5-Triazine-2,4,6-triamine-15N3
Empirical Formula (Hill Notation):
C3H615N3N3
CAS Number:
Molecular Weight:
129.10
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing