
Luteinizing hormone releasing hormone

Synonym(s):
LH-RH, [Gly-OH10]-LH-RH, pGlu-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly
Empirical Formula (Hill Notation):
C55H74N16O14
CAS Number:
Molecular Weight:
1183.27
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing