
Lithocholic acid

Synonym(s):
3α-Hydroxy-5β-cholan-24-oic acid, 3α-Hydroxy-5β-cholanic acid, 5β-Cholan-24-oic acid-3α-ol
Empirical Formula (Hill Notation):
C24H40O3
CAS Number:
Molecular Weight:
376.57
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Tissue
Functional Group
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3