Skip to Content
Merck
vibrant-m

Levalbuterol Related Compound C

vibrant-m
Synonym(s):
4-[2-(tert-Butylamino)-1-hydroxyethyl]-2-(methoxymethyl)phenol, α-{[(1,1-Dimethylethyl)amino]methyl}-4-hydroxy-3-(methoxymethyl)benzenemethanol, Albuterol monomethyl ether
Empirical Formula (Hill Notation):
C14H23NO3
CAS Number:
Molecular Weight:
253.34

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1