Skip to Content
Merck
Imidazole ≥99% (titration), free-flowing, Redi-Dri™

Imidazole

Imidazole ≥99% (titration), free-flowing, Redi-Dri™
Synonym(s):
1,3-Diaza-2,4-cyclopentadiene, Glyoxaline, Imidazole, Imidazole, ULTROL® Grade
Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08

Products

Product Category
Brand
Boiling Point (°C)
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique