Skip to Content
Merck
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice) ≥70% (HPLC)

Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice)

Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice) ≥70% (HPLC)
Synonym(s):
3-O-(2-O-β-D-Glucopyranuronosyl-α-D-glucopyranuronosyl)-18β-glycyrrhetinic acid ammonium salt, Glycyrrhizin
Linear Formula:
C42H62O16 · NH3
CAS Number:
Molecular Weight:
839.96
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer