
Glibenclamide impurity B

Synonym(s):
Methyl {{4-{2-[(5-chloro-2-methoxybenzoyl)amino]ethyl}phenyl}sulfonyl}carbamate
Empirical Formula (Hill Notation):
C18H19ClN2O6S
CAS Number:
Molecular Weight:
426.87
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing