
γ,γ-Dimethylallyl pyrophosphate ammonium salt

Synonym(s):
3-Methyl-2-butenyl pyrophosphate ammonium salt, DMAPP, Prenyl pyrophosphate ammonium salt
Empirical Formula (Hill Notation):
C5H12O7P2 · xNH3
CAS Number:
Molecular Weight:
246.09 (free acid basis)
Products
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing