
Gabapentin Related Compound A

Synonym(s):
4,4-Pentamethylene-2-pyrrolidinone, 3-Azaspiro[4.5]decan-2-one
Empirical Formula (Hill Notation):
C9H15NO
CAS Number:
Molecular Weight:
153.22
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2