
Flurprimidol

Synonym(s):
α-(1-Methylethyl)-α-[4-(trifluoromethoxy)phenyl]-5-pyrimidinemethanol
Empirical Formula (Hill Notation):
C15H15F3N2O2
CAS Number:
Molecular Weight:
312.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2