Skip to Content
Merck
Fluridon PESTANAL®, analytical standard

Fluridon

Fluridon PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C19H14F3NO
CAS Number:
Molecular Weight:
329.32
EC Number:

Products

Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing