
Flurbiprofen sodium

Synonym(s):
Flurbiprofen sodium salt
Empirical Formula (Hill Notation):
C15H12FNaO2
CAS Number:
Molecular Weight:
266.24
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2