
Fluocinolone acetonide

Synonym(s):
Fluocinolone acetonide, 6α,9α-Difluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, 6α,9α-Difluoro-16α-hydroxyprednisolone 16,17-acetonide, 6α-Fluorotriamcinolone acetonide, Sinalar, Synandone
Empirical Formula (Hill Notation):
C24H30F2O6
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing