
Fexofenadine hydrochloride

Synonym(s):
Fexofenidine hydrochloride, MDL 16455 hydrochloride, Terfenidine carboxylate hydrochloride
Empirical Formula (Hill Notation):
C32H39NO4 · HCl
CAS Number:
Molecular Weight:
538.12
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing