Skip to Content
BG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
EZ-Fit
®
Manifold Kit
Products
Product Category
manifolds (4)
Brand
Millipore (4)
Agency
Diameter (mm)
Application
Membrane Material
Pore size (µm)
Sterilization Method
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
MKHAWG474
EZ-Fit
®
Manifold Kit,
Contains 4 boxes of EZ-Pak™ membrane filters, EZ-Fit
®
3-head Manifold with filtration heads
Expand
View Pricing
KMEFHAW10S
EZ-Fit
®
Manifold Kit,
Contains 4 boxes of EZ-Fit™ 100 mL Funnels with single packaging, EZ-Fit
®
3-head Manifold with filtration heads
Expand
View Pricing
KMEFHAW10B
EZ-Fit
®
Manifold Kit,
Contains 4 boxes of EZ-Fit™ 100 mL Funnels with bulk packaging, EZ-Fit
®
3-head Manifold with filtration heads
Expand
View Pricing
KMEFHAW25B
EZ-Fit
®
Manifold Kit,
Contains 4 boxes of EZ-Fit™ 250 mL Funnels with bulk packaging, EZ-Fit
®
3-head Manifold with filtration heads
Expand
View Pricing