Skip to Content
Merck
Ethyl laurate analytical standard

Ethyl laurate

Ethyl laurate analytical standard
Synonym(s):
Ethyl dodecanoate, Ethyl laurate, Lauric acid ethyl ester
Linear Formula:
CH3(CH2)10COOC2H5
CAS Number:
Molecular Weight:
228.37
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing