
Eriochrome black T (C.I. 14645)

Synonym(s):
Eriochrome black T (C.I. 14645)
Empirical Formula (Hill Notation):
C20H12N3NaO7S
CAS Number:
Molecular Weight:
461.38
Products
Brand
Color
Formula Weight
Physical Form
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing