Skip to Content
Merck
(±)−α−Tocopherol (Vitamin E) solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

(±)−α−Tocopherol (Vitamin E) solution

(±)−α−Tocopherol (Vitamin E) solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
(±)-α-Tocopherol, DL-all-rac-α-Tocopherol, Vitamin E
Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique