
Diquat metabolite TOPPS

Synonym(s):
1-Oxo-1,2,3,4-tetrahydropyrido[1,2-a]pyrazin-5-ium bromide
Empirical Formula (Hill Notation):
C8H9BrN2O
CAS Number:
Molecular Weight:
229.07
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing