Skip to Content
Merck
Dibenz [b,f]azepine-5-carbonyl chloride 90%

Dibenz [b,f]azepine-5-carbonyl chloride

Dibenz [b,f]azepine-5-carbonyl chloride 90%
Empirical Formula (Hill Notation):
C15H10ClNO
CAS Number:
Molecular Weight:
255.70

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing