
Decursin

Synonym(s):
(+)-Decursin, (7S)-7,8-Dihydro-8,8-dimethyl-2-oxo-2H,6H-benzo[1,2-b:5,4-b′]dipyran-7-yl 3-methyl-2-butenoate, 3-Methyl-2-butenoic acid (7S)-7,8-dihydro-8,8-dimethyl-2-oxo-2H,6H-pyrano[3,2-g]-1-benzopyran-7-yl ester
Empirical Formula (Hill Notation):
C19H20O5
CAS Number:
Molecular Weight:
328.36
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing