
Cyflumetofen

Synonym(s):
2-Methoxyethyl (RS)-2-(4-tert-butylphenyl)-2-cyano-3-oxo-3-(α,α,α-trifluoro-o-tolyl)propionate
Empirical Formula (Hill Notation):
C24H24F3NO4
CAS Number:
Molecular Weight:
447.45
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2