
Cycloxidim

Empirical Formula (Hill Notation):
C17H27NO3S
CAS Number:
Molecular Weight:
325.47
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing