Clobazam
Synonym(s):
7-Chloro-1-methyl-5-phenyl-1H-1,5-benzodiazepine-2,4-(3H,5H)-dione
Empirical Formula (Hill Notation):
C16H13ClN2O2
CAS Number:
Molecular Weight:
300.74
Products
Product Category
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing