Skip to Content
BG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chromium Standard for ICP
Products
Brand
Supelco (3)
Product Line
Format
Chemical Composition
Element
Quality Segment
Application
Special Grade
certified reference material (3)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
68131
Chromium Standard for ICP,
Trace
CERT
®
, 1 g/L Cr in nitric acid (nominal concentration), prepared with (NH
4
)
2
Cr
2
O
7
Expand
View Pricing
74582
Chromium Standard for ICP,
Trace
CERT
®
, 1 g/L Cr in nitric acid (nominal concentration), prepared with Cr(NO
3
)
3
x 9H
2
O
Expand
View Pricing
93104
Chromium Standard for ICP,
Trace
CERT
®
, 10 g/L Cr in nitric acid (nominal concentration)
Expand
View Pricing