
Carbazole

Synonym(s):
Carbazole
Empirical Formula (Hill Notation):
C12H9N
CAS Number:
Molecular Weight:
167.21
Products
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing