
Boron trifluoride propanol complex

Linear Formula:
BF3 · CH3CH2CH2OH
CAS Number:
Molecular Weight:
127.90
Products
Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing